EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Pyrone derivatives from the endophytic fungus Alternaria tenuissima SP-07 of Chinese herbal medicine Salvia przewalskii.
PMID 25284429
Abstract
Three new pyrones, solanapyrones P-R (1-3), were afforded by the extracts of the endophytic fungus Alternaria tenuissima SP-07 isolated from the fresh root of Chinese herbal medicine Salvia przewalskii, along with the known solanapyrones (4-6) and benzopyrones (7-9). Solanapyrones P (1) and Q (2) possess an unprecedented nor-solanapyrone skeleton as natural products. Their structures were determined on the basis of NMR and HR-ESI-MS analysis. The plausible biosynthetic pathways to those unknown compounds were discussed. All the isolated compounds were evaluated for their antibacterial activities against six bacteria.