EMAIL THIS PAGE TO A FRIEND
Indian journal of pharmaceutical sciences
Relationship between Antifungal Activity against Candida albicans and Electron Parameters of Selected N-Heterocyclic Thioamides.
PMID 25284926
Abstract
Due to the increasing demand for new pharmaceuticals showing biological activity against pathogenic microorganisms, there is increasing search for new compounds with predicted biological activity. Variously substituted thioamide derivatives with 1.3 and 1.2 ring of thiazole and 1,3,4-thiadiazole, as well as pyrazole were assessed for their activity against Candida albicans. Activity of majority of tested thioamides was larger as compared with that of the reference drugs. The electron parameters of obtained N-heterocyclic thioamides were determined and dependencies on their biological activity against Candida albicans were studied. The best electron compliance of produced bindings with the activity against Candida albicans was observed for the derivatives containing 1,3,4-thiadiazole ring.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7000000
Itraconazole, European Pharmacopoeia (EP) Reference Standard
C35H38Cl2N8O4