EMAIL THIS PAGE TO A FRIEND
European journal of heart failure
Sex-dependent regulation of fibrosis and inflammation in human left ventricular remodelling under pressure overload.
PMID 25287281
Abstract
Women with aortic stenosis develop a more concentric form of LV hypertrophy than men. However, the molecular factors underlying sex differences in LV remodelling are incompletely understood. We took an unbiased approach to identify sex-specific patterns in gene expression and pathway regulation, and confirmed the most prominent findings in human hearts. Echocardiography was performed in 104 patients (53.8% women) with aortic stenosis before aortic valve replacement. LV mass, LV end-diastolic diameter, and relative wall thickness were included in a factor analysis to generate an index classifying LV remodelling as adaptive or maladaptive. Maladaptive remodelling was present in 64.6% of male and in 32.7% of female patients (P < 0.01). Genome-wide expression profiling of LV samples was performed in a representative subgroup of 19 patients (52.6% women) compared with samples from healthy controls (n = 18). Transcriptome characterization revealed that fibrosis-related genes/pathways were induced in male overloaded ventricles, while extracellular matrix-related and inflammatory genes/pathways were repressed in female overloaded ventricles (adjusted P < 0.05). We confirmed gene regulation by quantitative real-time reverse transcription-polymerase chain reaction and immunoblotting analysis, and we further demonstrate the relevance of our findings by histological documentation of higher fibrosis in men than in women. We conclude that in pressure overload distinct molecular processes are regulated between men and women. Maladaptive LV remodelling occurs more frequently in men and is associated with greater activation of profibrotic and inflammatory markers. Collectively, sex-specific regulation of these processes may contribute to sex differences in the progression to heart failure.