EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Gastroprotective effects of L-lysine salification of ketoprofen in ethanol-injured gastric mucosa.
PMID 25287669
Abstract
Ketoprofen L-lysine salt (KLS), a NSAID, is widely used for its analgesic efficacy and tolerability. L-lysine salification was reported to increase the solubility and the gastric absorption and tolerance of ketoprofen. Since the management of NSAIDs gastrotoxicity still represents a major limitation in prolonged therapies, mainly when gastric lesions are present, this study investigated the gastro-protective activity of L-lysine by using a well-established model of gastric mucosa injury, the ethanol-gastric injury model. Several evidences show that the damaging action of ethanol could be attributed to the increase of ROS, which plays a key role in the increase of lipid peroxidation products, including malonyldialdehyde and 4-hydroxy-2-nonenal. With the aim to unravel the mechanism of L-lysine gastroprotection, cellular MDA levels and 4-HNE protein adducts as markers of lipid peroxidation and a panel of key endogenous gastro-protective proteins were assayed. The data obtained indicate a gastroprotective effect of L-lysine on gastric mucosa integrity.