EMAIL THIS PAGE TO A FRIEND
Pharmaceutical biology
Lipidomics investigation of reversal effect of glycyrrhizin (GL) towards lithocholic acid (LCA)-induced alteration of phospholipid profiles.
PMID 25289528
Abstract
Glycyrrhizin (GL), the major ingredient isolated from licorice, exerts multiple pharmacological activities. To elucidate the protective mechanism of GL towards lithocholic acid (LCA)-induced liver toxicity using lipidomics. GL (200 mg/kg) dissolved in corn oil was treated intraperitoneally for 7 d. On the 4th day, 200 mg/kg LCA was used to treat mice (i.p., twice daily) for another 4 d. The protective role of GL towards LCA-induced liver toxicity was investigated through evaluating the liver histology and the activity of alanine transaminase (ALT). The complete lipid profile was employed using UFLC-Triple TOF MS-based lipidomics. Intraperitoneal (i.p.) administration of 200 mg/kg GL can significantly protect LCA-induced liver damage, indicated by alleviated histology alteration and prevention of the ALT elevation. Lipidomics analysis can well separate the control group from LCA-treated group, and three lipid components were major contributors, including LPC 16:0, LPC 18:0, and LPC 18:2. GL treatment can significantly prevent LCA-induced reduction of these three lipid compounds, providing a new explanation for GL's protection mechanism towards LCA-induced liver toxicity. The recent study highlights the importance of lipidomics in elucidating the therapeutic mechanism of herbs.
Related Materials