EMAIL THIS PAGE TO A FRIEND
Biopreservation and biobanking
Method validation for preparing urine samples for downstream proteomic and metabolomic applications.
PMID 25289566
Abstract
Formal validation of methods for biospecimen processing in the context of accreditation in laboratories and biobanks is lacking. A protocol for processing of a biospecimen (urine) was validated for fitness-for-purpose in terms of key downstream endpoints. Urine processing was optimized for centrifugation conditions on the basis of microparticle counts at room temperature (RT) and at 4°C. The optimal protocol was validated for performance (microparticle counts), and for reproducibility and robustness for centrifugation temperature (4°C vs. RT) and brake speed (soft, medium, hard). Acceptance criteria were based on microparticle counts, cystatin C and creatinine concentrations, and the metabolomic profile. The optimal protocol was a 20-min, 12,000 g centrifugation at 4°C, and was validated for urine collection in terms of microparticle counts. All reproducibility acceptance criteria were met. The protocol was robust for centrifugation at 4°C versus RT for all parameters. The protocol was considered robust overall in terms of brake speeds, although a hard brake gave significantly fewer microparticles than a soft brake. We validated a urine processing method suitable for downstream proteomic and metabolomic applications. Temperature and brake speed can influence analytic results, with 4°C and high brake speed considered optimal. Laboratories and biobanks should ensure these conditions are systematically recorded in the scope of accreditation.