EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Macrophages sense and kill bacteria through carbon monoxide-dependent inflammasome activation.
PMID 25295542
Abstract
Microbial clearance by eukaryotes relies on complex and coordinated processes that remain poorly understood. The gasotransmitter carbon monoxide (CO) is generated by the stress-responsive enzyme heme oxygenase-1 (HO-1, encoded by Hmox1), which is highly induced in macrophages in response to bacterial infection. HO-1 deficiency results in inadequate pathogen clearance, exaggerated tissue damage, and increased mortality. Here, we determined that macrophage-generated CO promotes ATP production and release by bacteria, which then activates the Nacht, LRR, and PYD domains-containing protein 3 (NALP3) inflammasome, intensifying bacterial killing. Bacterial killing defects in HO-1-deficient murine macrophages were restored by administration of CO. Moreover, increased CO levels enhanced the bacterial clearance capacity of human macrophages and WT murine macrophages. CO-dependent bacterial clearance required the NALP3 inflammasome, as CO did not increase bacterial killing in macrophages isolated from NALP3-deficient or caspase-1-deficient mice. IL-1β cleavage and secretion were impaired in HO-1-deficient macrophages, and CO-dependent processing of IL-1β required the presence of bacteria-derived ATP. We found that bacteria remained viable to generate and release ATP in response to CO. The ATP then bound to macrophage nucleotide P2 receptors, resulting in activation of the NALP3/IL-1β inflammasome to amplify bacterial phagocytosis by macrophages. Taken together, our results indicate that macrophage-derived CO permits efficient and coordinated regulation of the host innate response to invading microbes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405949
Anti-HMOX1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000635
Anti-HMOX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101053
Anti-HMOX1 antibody produced in rabbit, affinity isolated antibody
SAB1410641
Anti-HMOX1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501287
Anti-P2RX7 (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV35518
Anti-P2RX7 antibody produced in rabbit, IgG fraction of antiserum
SAB2101706
Anti-P2RX7 antibody produced in rabbit, affinity isolated antibody
SAB2104209
Anti-P2RX7 antibody produced in rabbit, affinity isolated antibody
HPA034968
Anti-P2RX7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044141 Anti-P2RX7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
P9122
Anti-Purinergic Receptor P2X7 (extracellular) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2500916
Anti-SDHB antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002867
Anti-SDHB antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102103
Anti-SDHB antibody produced in rabbit, affinity isolated antibody
HPA002868
Anti-SDHB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106796
Anti-SDHB antibody produced in rabbit, affinity isolated antibody
SAB1411242
Anti-SDHB antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0003162M1
Monoclonal Anti-HMOX1 antibody produced in mouse, clone 5C6, purified immunoglobulin, buffered aqueous solution
AMAB90708
Monoclonal Anti-SDHB antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0349, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90706
Monoclonal Anti-SDHB antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0347, purified immunoglobulin, buffered aqueous glycerol solution