EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Endosomal localization of TLR8 confers distinctive proteolytic processing on human myeloid cells.
PMID 25297876
Abstract
Nucleic acid-sensing TLRs are involved in both antimicrobial immune responses and autoimmune inflammation. TLR8 is phylogenetically and structurally related to TLR7 and TLR9, which undergo proteolytic processing in the endolysosomes to generate functional receptors. Recent structural analyses of human TLR8 ectodomain and its liganded form demonstrated that TLR8 is also cleaved, and both the N- and C-terminal halves contribute to ligand binding. However, the structures and ssRNA recognition mode of endogenous TLR8 in human primary cells are largely unknown. In this study, we show that proteolytic processing of TLR8 occurs in human monocytes and macrophages in a different manner compared with TLR7/9 cleavage. The insertion loop between leucine-rich repeats 14 and 15 in TLR8 is indispensable for the cleavage and stepwise processing that occurs in the N-terminal fragment. Both furin-like proprotein convertase and cathepsins contribute to TLR8 cleavage in the early/late endosomes. TLR8 recognizes viral ssRNA and endogenous RNA, such as microRNAs, resulting in the production of proinflammatory cytokines. Hence, localization sites of the receptors are crucial for the nucleic acid-sensing mode and downstream signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA001012
Anti-ARHGAP4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001083
Anti-ARHGAP4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410749
Anti-ARHGAP4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501291
Anti-CANX antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA021178
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA021230
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA021799
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
HPA002877
Anti-PGRMC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101782
Anti-PGRMC1 antibody produced in rabbit, affinity isolated antibody
HPA001608
Anti-TLR8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500307
Anti-TLR8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500768
Anti-TLR8 antibody produced in rabbit, affinity isolated antibody
SAB3500769
Anti-TLR8 antibody produced in rabbit, affinity isolated antibody
63580
D-(+)-Mannose, for microbiology, ≥99%
C6H12O6
63582
D-(+)-Mannose, for microbiology, ≥99.0% (sum of enantiomers, HPLC)
C6H12O6
EHU093811 MISSION® esiRNA, esiRNA human TLR8 (esiRNA1)
EMU079161 MISSION® esiRNA, esiRNA targeting mouse Tlr8 (esiRNA1)
WH0000393M1
Monoclonal Anti-ARHGAP4 antibody produced in mouse, clone 3F7, purified immunoglobulin, buffered aqueous solution
SAB1412794
Monoclonal Anti-PGRMC1 antibody produced in mouse, clone 3F7, purified immunoglobulin, 1 mg/mL