EMAIL THIS PAGE TO A FRIEND
Journal of medical microbiology
Selective growth-inhibitory effect of 8-hydroxyquinoline towards Clostridium difficile and Bifidobacterium longum subsp. longum in co-culture analysed by flow cytometry.
PMID 25298160
Abstract
The major risk factor for Clostridium difficile infection (CDI) is the use of antibiotics owing to the disruption of the equilibrium of the host gut microbiota. To preserve the beneficial resident probiotic bacteria during infection treatment, the use of molecules with selective antibacterial activity enhances the efficacy by selectively removing C. difficile. One of them is the plant alkaloid 8-hydroxyquinoline (8HQ), which has been shown to selectively inhibit clostridia without repressing bifidobacteria. Selective antimicrobial activity is generally tested by culture techniques of individual bacterial strains. However, the main limitation of these techniques is the inability to describe differential growth dynamics of more bacterial strains in co-culture within the same experiment. In the present study, we combined fluorescent in situ hybridization and flow cytometry to describe the changes in active and non-active cells of a mixed culture formed by the opportunistic pathogen C. difficile CECT 531 and the beneficial Bifidobacterium longum subsp. longum CCMDMND BL1 after exposure to 8HQ. It was observed that without 8HQ, the proportion of both strains was almost equal, oscillating between 22.7 and 77.9 % during a time lapse of 12 h, whereas with 8HQ the proportion of active C. difficile decreased after 4 h, and persisted only between 8.8 and 17.5 %. In contrast, bifidobacterial growth was not disturbed by 8HQ. The results of this study showed the selective inhibitory effect of 8HQ on clostridial and bifidobacterial growth dynamics, and the potential of this compound for the development of selective agents to control CDIs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36524
8-Quinolinol, PESTANAL®, analytical standard
C9H7NO
55100
8-Quinolinol hemisulfate salt, ≥98.0% (dry substance, T), yellow
C18H14N2O2 · H2SO4
C1276
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
C3H7NO2S · HCl
C7477
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
C3H7NO2S · HCl
30120
L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)
C3H7NO2S · HCl
W778567
L-Cysteine hydrochloride, FG, product of Wacker Chemie AG
C3H7NO2S · HCl
P9172
Pyronin Y, for NA electrophoresis
C17H19ClN2O
83200
Pyronin Y, for microscopy (Bot., Fl., Hist.)
C17H19ClN2O
213519
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
C17H19ClN2O
T3383
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
C22H24N2O8 · HCl
T7660
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
C22H24N2O8 · HCl
T8032
Tetracycline hydrochloride, powder
C22H24N2O8 · HCl
T4062
Tetracycline hydrochloride, meets USP testing specifications
C22H24N2O8 · HCl
31741
Tetracycline hydrochloride, VETRANAL, analytical standard
C22H24N2O8 · HCl
PHR1041
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C22H24N2O8 · HCl
1651009
Tetracycline hydrochloride, United States Pharmacopeia (USP) Reference Standard
C22H24N2O8 · HCl