EMAIL THIS PAGE TO A FRIEND
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
The progesterone and estrogen modify the uterine prolactin and prolactin receptor expression of hyperprolactinemic mice.
PMID 25299230
Abstract
The aim of this study was to evaluate the effects of metoclopramide-induced hyperprolactinemia on the prolactin (PRL) and PRL receptor's expression in the uterus of mice. For this purpose, 49 Swiss mice were divided into the following groups: GrSS (non-ovariectomized mice given vehicle); GrMET (non-ovariectomized mice treated with metoclopramide); OvSS (ovariectomized mice given vehicle); OvMET (ovariectomized mice treated with metoclopramide); OvMET+17βE (ovariectomized mice treated with metoclopramide and 17β estradiol); OvMET+MP (ovariectomized mice treated with metoclopramide and micronized progesterone); OvMET+17βE+MP (ovariectomized mice treated with metoclopramide and a solution of 17β estradiol and micronized progesterone). Immunohistochemical analyzes were evaluated semi-quantitatively. Our results showed that GrMET, OvMET+MP, and OvMET+17βE+MP presented strong PRL expression. OvMET and OvMET+17βE presented mild reaction, while GrSS and OvSS presented weak reaction. Concerning PRL receptor, OvMET+MP and OvMET+17βE+MP showed strong reaction; GrMET, OvSS, and OvMET+17βE showed mild reaction; and GrSS and OvMET showed weak reaction. These findings suggest that progesterone alone or in combination with estrogen may increase the expression of uterine PRL and PRL receptor.
Related Materials