EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Arsenic removal from drinking water by a household sand filter in Vietnam--effect of filter usage practices on arsenic removal efficiency and microbiological water quality.
PMID 25300017
Abstract
Household sand filters are applied to treat arsenic- and iron-containing anoxic groundwater that is used as drinking water in rural areas of North Vietnam. These filters immobilize poisonous arsenic (As) via co-oxidation with Fe(II) and sorption to or co-precipitation with the formed Fe(III) (oxyhydr)oxides. However, information is lacking regarding the effect of the frequency and duration of filter use as well as of filter sand replacement on the residual As concentrations in the filtered water and on the presence of potentially pathogenic bacteria in the filtered and stored water. We therefore scrutinized a household sand filter with respect to As removal efficiency and the presence of fecal indicator bacteria in treated water as a function of filter operation before and after sand replacement. Quantification of As in the filtered water showed that periods of intense daily use followed by periods of non-use and even sand replacement did not significantly (p<0.05) affect As removal efficiency. The As concentration was reduced during filtration from 115.1 ± 3.4 μg L(-1) in the groundwater to 5.3 ± 0.7 μg L(-1) in the filtered water (95% removal). The first flush of water from the filter contained As concentrations below the drinking water limit and suggests that this water can be used without risk for human health. Colony forming units (CFUs) of coliform bacteria increased during filtration and storage from 5 ± 4 per 100mL in the groundwater to 5.1 ± 1.5 × 10(3) and 15 ± 1.4 × 10(3) per 100mL in the filtered water and in the water from the storage tank, respectively. After filter sand replacement, CFUs of Escherichia coli of <100 per 100mL were quantified. None of the samples contained CFUs of Enterococcus spp. No critical enrichment of fecal indicator bacteria belonging to E. coli or Enterococcus spp. was observed in the treated drinking water by qPCR targeting the 23S rRNA gene. The results demonstrate the efficient and reliable performance of household sand filters regarding As removal, but indicate a potential risk for human health arising from the enrichment of coliform bacteria during filtration and from E. coli cells that are introduced by sand replacement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
421553
Silica
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
560979
Silica, mesostructured, MSU-F (cellular foam)
O2Si
541036
Silica, mesostructured, HMS (wormhole)
O2Si
643637
Silica, mesostructured, MSU-H (large pore 2D hexagonal)
O2Si
643645
Silica, mesostructured, MCM-41 type (hexagonal)
O2Si
637238
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
O2Si
637246
Silicon dioxide, nanopowder (spherical, porous), 5-15 nm particle size (TEM), 99.5% trace metals basis
O2Si