EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Effects of iron supplementation on blood adenine deaminase activity and oxidative stress in Trypanosoma evansi infection of rats.
PMID 25300765
Abstract
The aim of this study was to assess the effects of iron supplementation on oxidative stress and on the activity of the adenosine deaminase (ADA) in rats experimentally infected by Trypanosoma evansi. For this purpose, 20 rats were divided into four experimental groups with five animals each as follows: groups A and B were composed by healthy animals, while animals from groups C and D were infected by T. evansi. Additionally, groups B and D received two subcutaneous doses of iron (60 mg kg(-1)) within an interval of 5 days. Blood samples were drawn on day 8 post infection in order to assess hematological and biochemical variables. Among the main results are: (1) animals from group C showed reduced erythrogram (with tendency to anemia); however the same results were not observed for group D; this might be a direct effect of free iron on trypanosomes which helped to reduce the parasitemia and the damage to erythrocytes caused by the infection; (2) iron supplementation was able to reduce NOx levels by inhibiting iNOS, and thus, providing an antioxidant action and, indirectly, reducing the ALT levels in groups Band D; (3) increase FRAP levels in group D; (4) reduce ADA activity in serum and erythrocytes in group C; however, this supplementation (5) increased the protein oxidation in groups B and D, as well as group C (positive control). Therefore, iron showed antioxidant and oxidant effects on animals that received supplementation; and it maintained the activity of E-ADA stable in infected/supplemented animals.