EMAIL THIS PAGE TO A FRIEND
BMC cancer
CUB domain containing protein 1 (CDCP1) modulates adhesion and motility in colon cancer cells.
PMID 25301083
Abstract
Deregulated expression of the transmembrane glycoprotein CDCP1 (CUB domain-containing protein-1) has been detected in several cancers including colon, lung, gastric, breast, and pancreatic carcinomas. CDCP1 has been proposed to either positively or negatively regulate tumour metastasis. In this study we assessed the role of CDCP1 in properties of cells that are directly relevant to metastasis, namely adhesion and motility. In addition, association between CDCP1 and the tetraspanin protein CD9 was investigated. CDCP1 and CD9 protein expression was measured in a series of colon cancer cell lines by flow cytometry and Western blotting. Adhesion of Colo320 and SW480 cells was determined using a Matrigel adhesion assay. The chemotactic motility of SW480 cells in which CDCP1 expression had been reduced by RNA interference was analysed using the xCELLigence system Real-Time Cell Analyzer Dual Plates combined with 8 μm pore filters. Detergent-resistant membrane fractions were generated following density gradient centrifugation and the CDCP1 and CD9 protein composition of these fractions was determined by Western blotting. The potential association of the CDCP1 and CD9 proteins was assessed by co-immunoprecipitation. Engineered CDCP1 expression in Colo320 cells resulted in a reduction in cell adhesion to Matrigel. Treatment of SW480 cells with CDCP1 siRNA reduced serum-induced chemotaxis. CDCP1 and CD9 cell-surface protein and mRNA levels showed a positive correlation in colon cancer cell lines and the proteins formed a low-level, but detectable complex as judged by co-sedimentation of detergent lysates of HT-29 cells in sucrose gradients as well as by co-immunoprecipitation in SW480 cell lysates. A number of recent studies have assigned a potentially important role for the cell-surface protein CDCP1 in invasion and metastasis of a several types of human cancer cells. In this study, CDCP1 was shown to modulate cell-substratum adhesion and motility in colon cancer cell lines, with some variation depending on the colon cancer cell type. CDCP1 and CD9 were co-expressed at the mRNA and protein level and we obtained evidence for the presence of a molecular complex of these proteins in SW480 colon cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1407905
Anti-CDCP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100532
Anti-DBI antibody produced in rabbit, affinity isolated antibody
SAB1410285
Anti-DBI antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA061260
Anti-HPS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
B3773 Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
B5283 Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
A1949 Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
P5884 Brij® 58, average Mn ~1124
P6136
Brij® O10
C38H76O11
SAB4700515 Monoclonal Anti-CD71 antibody produced in mouse, clone MEM-75, purified immunoglobulin, buffered aqueous solution
SAB4700520 Monoclonal Anti-CD71 antibody produced in mouse, clone MEM-189, purified immunoglobulin, buffered aqueous solution
SAB4700092 Monoclonal Anti-CD9 antibody produced in mouse, clone MEM-61, purified immunoglobulin, buffered aqueous solution
SAB4700618 Monoclonal Anti-CD9 antibody produced in mouse, clone IVA50, purified immunoglobulin, buffered aqueous solution
SAB1402143
Monoclonal Anti-CD9 antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
SAB4100431
Monoclonal Anti-CDCP1 antibody produced in mouse, clone CDCP1-A364, culture supernatant
SAB1403056
Monoclonal Anti-FLOT1 antibody produced in mouse, clone 4A1, ascites fluid
G2904 Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
A8062 Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148 Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891 Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
A9452 Monoclonal Anti-Goat/Sheep IgG–Peroxidase antibody produced in mouse, clone GT-34, purified immunoglobulin, lyophilized powder
I6135 Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
I6260 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
I6760 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
F5016 Monoclonal Anti-Human IgG (Fc specific)−FITC antibody produced in mouse, clone HP-6017, purified immunoglobulin, buffered aqueous solution
I5885 Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
I9513 Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
I9888 Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
R1008 Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
A2556 Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution