EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Acacetin blocks kv1.3 channels and inhibits human T cell activation.
PMID 25301362
Abstract
Acacetin, a natural flavonoid compound, has been proven to exert anti-inflammatory and immunomodulatory effects. Kv1.3 channels, highly expressed in human T cells, are attractive therapeutic targets to treat inflammatory and immunological disorders. The present study was designed to characterize the inhibition of Kv1.3 channels by Acacetin in human T cells and examine its role in T cell activation. Whole-cell patch-clamp was applied to record the Kv1.3 and KCa currents in human T cells; Western blot was used to detect Kv1.3 expression as well as NFAT1 and NF-κB activity; Fluo-4, CCK-8 and an ELISA kit were used to measure Ca(2+) influx, proliferation, and IL-2 secretion, respectively. Acacetin decreased the Kv1.3 current, accelerated the decay rate and negatively shifted the steady-state inactivation curves in a concentration-dependent manner. The IC50 values at +40 mV for peak and the current at end of pulse were 21.09 ± 2.75 and 3.63 ± 0.25 μmol/L, respectively. Treatment with Acacetin for 24 h significantly inhibited Kv1.3 protein expression. Additionally, paralleling Kv1.3 inhibition, Acacetin also inhibited Ca(2+) influx, the Ca(2+)-activated transcription factors NFAT1, NF-κB p65/p50 activity, and proliferation as well as IL-2 production. Small interfering RNA against Kv1.3 reduced the inhibitory effect of Acacetin on IL-2 secretion. Acacetin blocks the Kv1.3 channel and inhibits human T cell activation. This action most likely contributes to its immunomodulatory and anti-inflammatory actions.