EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Litorivivens lipolytica gen. nov., sp. nov., a lipolytic bacterium isolated from a tidal flat.
PMID 25301540
Abstract
A Gram-stain-negative, flagellated, aerobic and rod-shaped or ovoid bacterial strain, designated HJTF-7(T), was isolated from a tidal flat on the South Sea of South Korea, and its taxonomic position was investigated using a polyphasic approach. Strain HJTF-7(T) grew optimally at 25 °C, at pH 7.0-8.0 and in the presence of 2.0% (w/v) NaCl. Neighbour-joining, maximum-likelihood and maximum-parsimony phylogenetic trees, based on 16S rRNA gene sequences, showed that strain HJTF-7(T) joined the cluster comprising the type strains of species of the genera Spongiibacter and Zhongshania. Strain HJTF-7(T) exhibited 16S rRNA gene sequence similarities of 90.4-92.5% to the type strains of species of the genera Spongiibacter and Zhongshania and of less than 91.5% to the type strains of other recognized species. Strain HJTF-7(T) contained Q-8 as the predominant ubiquinone. The major fatty acids were iso-C17:1ω9c, iso-C15:0, iso-C17:0, iso-C11:0 3-OH and C17:1ω8c and the major polar lipids were phosphatidylethanolamine and phosphatidylglycerol. The fatty acid and polar lipid profiles of strain HJTF-7(T) were distinct from those of members of the genera Spongiibacter and Zhongshania. The DNA G+C content of strain HJTF-7(T) was 55.9 mol%. The phylogenetic data and differential chemotaxonomic and other phenotypic properties revealed that strain HJTF-7(T) represents a novel genus and species within the class Gammaproteobacteria, for which the name Litorivivens lipolytica gen. nov., sp. nov. is proposed. The type strain of Litorivivens lipolytica is HJTF-7(T) ( =KCTC 42157(T) =CECT 8654(T)).