EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Exhaust and evaporative emissions from motorcycles fueled with ethanol gasoline blends.
PMID 25302450
Abstract
The emission characteristics of motorcycles using gasoline and E10 (90% gasoline and 10% ethanol by volume) were investigated in this article. Exhaust and evaporative emissions of three motorcycles were investigated on the chassis dynamometer over the Urban Driving Cycle (UDC) and in the Sealed Housing for Evaporative Determination (SHED) including regulated and unregulated emissions. The regulated emissions were detected by an exhaust gas analyzer directly. The unregulated emissions including carbonyls and volatile organic compounds (VOCs) were sampled through battery-operated air pumps using tubes coated with 2,4-dinitrophenylhydrazine (DNPH) and Tenax TA, respectively. The experimental results showed that the emission factors of total hydrocarbons (THC) and carbon monoxide (CO) from E10 fueling motorcycles decreased by 26%-45% and 63%-73%, while the emission factor of NOx increased by 36%-54% compared with those from gasoline fueling motorcycles. For unregulated emissions, the emission amount of VOCs from motorcycles fueled with E10 decreased by 18%-31% while total carbonyls were 2.6-4.5 times higher than those for gasoline. For evaporative emissions of THC and VOCs, for gasoline or E10, the diurnal breathing loss (DBL) was higher than hot soak loss (HSL). Using E10 as a fuel does not make much difference in the amount of evaporative THC, while resulted in a slightly growth of 14%-17% for evaporative BETX (benzene, toluene, ethylbenzene, xylene).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
270709
Benzene, for HPLC, ≥99.9%
C6H6
32212
Benzene, puriss. p.a., Reag. Ph. Eur., ≥99.7%
C6H6
319953
Benzene, ACS reagent, ≥99.0%
C6H6
CRM48617 Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
C6H6
295116
Carbon monoxide, ≥99.0%
CO
24102
Ethanol, absolute, ≥99.8% (GC)
C2H6O
24105
Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%
C2H6O
24194
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
C2H6O
E-034
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-040
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material
C2H6O
E-031
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-038
Ethanol-100 (5 ampules/kit), 100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-041
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-056
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-043
Ethanol-20 (5 ampules/kit), 20 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-032
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-039
Ethanol-200 (5 ampules/kit), 200 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-035
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-033
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-045
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-036
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-044
Ethanol-400 (5 ampules/kit), 400 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-029
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-053
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-030
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-037
Ethanol-80, 80 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
459844
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
C2H6O
E7023
Ethyl alcohol, Pure, 200 proof, for molecular biology
C2H6O
493546
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
C2H6O
459828
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
C2H6O