EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Intramolecular metalloamination of N,N-dimethylhydrazinoalkenes: a versatile method to access functionalized piperidines and pyrrolidines.
PMID 25302713
Abstract
Highly diastereoselective metalloamination/cyclization reactions of zinc(II) hydrazides obtained through reaction of diethylzinc with N,N-dimethylhydrazinoalkenes are described. The resulting organozinc intermediates undergo facile allylation and acylation, in situ, to provide the corresponding functionalized piperidines and pyrrolidines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS003174
4-Hydroxypyrrolidine-2-carboxylic acid, AldrichCPR
C5H9NO3