EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Decreased N(6)-methyladenosine in peripheral blood RNA from diabetic patients is associated with FTO expression rather than ALKBH5.
PMID 25303482
Abstract
N(6)-methyladenosine (m(6)A) modification plays a fundamental role in the epigenetic regulation of the mammalian transcriptome. m(6)A can be demethylated by fat mass- and obesity-associated (FTO) protein and α-ketoglutarate-dependent dioxygenase alkB homolog 5 (ALKBH5) protein. However, the importance of m(6)A alteration in type 2 diabetes mellitus (T2DM) has not been explored. The objective of the study was to investigate whether m(6)A content was reduced in T2DM patients and whether m(6)A content was correlated with the mRNA expression levels of the FTO and ALKBH5 genes. In this case-control study, peripheral blood samples were obtained from 88 T2DM patients and 92 healthy controls. For the diabetic animal model experiment, blood samples were obtained from seven diabetic and eight nondiabetic rats. A sensitive liquid chromatography-electrospray ionization-tandem mass spectrometry method was developed for the determination of the m(6)A content in RNA, quantitative real-time PCR was used to examine the mRNA expression levels of the FTO and ALKBH5 genes, and high-resolution melting and DNA sequencing were used to detect FTO single-nucleotide polymorphisms. Our results showed that the m(6)A contents in the RNA from T2DM patients and diabetic rats were significantly lower compared with the control groups (P = 2.6 × 10(-24) for T2DM patients; P = .001 for diabetic rats, respectively), and T2DM can be characterized by the content of m(6)A. The mRNA expression level of FTO was significantly higher in T2DM patients than that of the controls (P = .0007) and was associated with the risk of T2DM (odds ratio 2.797, 95% confidence interval 1.452-5.389, P = .002). Moreover, the m(6)A contents were correlated with FTO mRNA expression. These data suggest that the increased mRNA expression of FTO could be responsible for the reduction of m(6)A in T2DM, which may further increase the risk of complications of T2DM. Low m(6)A should be investigated further as a novel potential biomarker of T2DM.