EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Metabolic profiling of Vitex agnus castus leaves, fruits and sprouts: analysis by LC/ESI/(QqQ)MS and (HR) LC/ESI/(Orbitrap)/MS n.
PMID 25305595
Abstract
Food supplements based on Vitex agnus castus L. (Verbenaceae) fruits, also known as chasteberry, are routinely used by women against somatic and psychic premenstrual symptoms such as depression, sadness or irritability. With the aim of highlighting the differences in the chemical profiles of cultivated fruits and different parts of wild plants (fruits, leaves and sprouts) of V. agnus castus, a method concerning with the quali-quantitative study of the derived hydroalcoholic extracts was carried out by using high-performance liquid chromatography coupled to electrospray negative ionization Orbitrap multicollisional high resolution mass spectrometry (LC/ESI/(Orbitrap)MS(n)) and high-performance liquid chromatography coupled to electrospray negative ionization triple quadrupole tandem mass spectrometry (LC/ESI/(QqQ)MS) in multiple reaction monitoring (MRM) mode.