EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
The protective role of ferulic acid on sepsis-induced oxidative damage in Wistar albino rats.
PMID 25305738
Abstract
Oxidative stress has an important role in the development of sepsis-induced multiorgan failure. Ferulic acid (FA), a well-established natural antioxidant, has several pharmacological activities including anti-inflammatory, anticancer and hepatoprotective. This study aimed to investigate the effects of FA on sepsis-induced oxidative damage in Wistar albino rats. Sepsis-induced DNA damage in the lymphocytes, liver and kidney cells of rats were evaluated by comet assay with and without formamidopyrimidine DNA glycosylase (Fpg). The oxidative stress parameters such as superoxide dismutase (SOD) and glutathione peroxidase (GSH-Px) activities and total glutathione (GSH) and malondialdehyde (MDA) levels were also measured. It is found that DNA damage in sepsis+FA-treated group was significantly lower than the sepsis group. FA treatment also decreased the MDA levels and increased the GSH levels and SOD and GSH-Px activities in the sepsis-induced rats. It seems that FA might have ameliorative effects against sepsis-induced oxidative damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E5134
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, for molecular biology, 99.0-101.0% (titration)
C10H14N2Na2O8 · 2H2O
E1644
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E6635
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, plant cell culture tested, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E5513
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E4884
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
ED2SS
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
C10H14N2Na2O8 · 2H2O
03677
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
C10H14N2Na2O8 · 2H2O
27285
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
03685
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
C10H14N2Na2O8 · 2H2O
2854
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
C10H14N2Na2O8
E7889
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
C10H14N2Na2O8