EMAIL THIS PAGE TO A FRIEND
Food chemistry
Direct detection of free fatty acids in edible oils using supercritical fluid chromatography coupled with mass spectrometry.
PMID 25306372
Abstract
Determination of free fatty acids (FFAs) in food products is of enormous interest mainly because they are related to the quality and authenticity of the oils. In this study, supercritical fluid chromatography (SFC), followed by an electrospray ionisation triple-quadrupole mass spectrometry (ESI-MS), is shown to provide a novel method for the separation and detection of FFAs in edible oils without any pretreatment. Eight FFAs were separated on a HSS C18 SB column with gradient elution within 3 min. Effects of different columns, modifiers and column temperature changes were evaluated. The results indicated the feasibility of this method for the high-throughput determination of individual FFAs with satisfactory correlation coefficients (R(2)>0.994) and good reproducibility of RSD<13.5% (intraday) and <15.0% (interday). By combined with principal component analysis (PCA), different types of edible oil were successfully distinguished into several categories, showing a potential application for the determination of oil quality or authenticity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3631
Arachidic acid, ≥99%
C20H40O2
10930
Arachidic acid, synthetic, ≥99.0% (GC)
C20H40O2
39383
Arachidic acid, analytical standard
C20H40O2
62230
Linoleic acid, analytical standard
C18H32O2
L2376
Linolenic acid, ≥99%
C18H30O2
62170
Linolenic acid, ~70% (GC)
C18H30O2
62160
Linolenic acid, analytical standard
C18H30O2
77402
Melting Point Standard 69-71°C, analytical standard
C18H36O2
M3128
Myristic acid, Sigma Grade, ≥99%
C14H28O2
70079
Myristic acid, analytical standard
C14H28O2
70082
Myristic acid, ≥98.0% (GC)
C14H28O2
W276405
Myristic acid, ≥95%, FCC, FG
C14H28O2
W276413
Myristic acid, natural
C14H28O2
130087
Myristic acid, 95%
C14H28O2
PHR1124
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C14H28O2
43051
Palmitic acid, certified reference material, TraceCERT®
C16H32O2
1492007
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
C16H32O2
76119
Palmitic acid, analytical standard
C16H32O2
PHR1120
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C16H32O2
P0090000
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
C16H32O2
76137
Stearic acid, certified reference material, TraceCERT®
C18H36O2
85679
Stearic acid, analytical standard
C18H36O2
85680
Stearic acid, puriss., ≥98.5% (GC)
C18H36O2
1621008
Stearic acid, United States Pharmacopeia (USP) Reference Standard
C18H36O2
S1340000
Stearic acid, European Pharmacopoeia (EP) Reference Standard
C18H36O2
PHR1114
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
C18H36O2