EMAIL THIS PAGE TO A FRIEND
Food chemistry
Nine novel angiotensin I-converting enzyme (ACE) inhibitory peptides from cuttlefish (Sepia officinalis) muscle protein hydrolysates and antihypertensive effect of the potent active peptide in spontaneously hypertensive rats.
PMID 25306378
Abstract
This study aimed to identify novel ACE inhibitory peptides from the muscle of cuttlefish. Proteins were hydrolyzed and the hydrolysates were then subjected to various types of chromatography to isolate the active peptides. Nine ACE inhibitory peptides were isolated and their molecular masses and amino acid sequences were determined using ESI-MS and ESI-MS/MS, respectively. The structures of the most potent peptides were identified as Val-Glu-Leu-Tyr-Pro, Ala-Phe-Val-Gly-Tyr-Val-Leu-Pro and Glu-Lys-Ser-Tyr-Glu-Leu-Pro. The first peptide displayed the highest ACE inhibitory activity with an IC50 of 5.22μM. Lineweaver-Burk plots suggest that Val-Glu-Leu-Tyr-Pro acts as a non-competitive inhibitor against ACE. Furthermore, antihypertensive effects in spontaneously hypertensive rats (SHR) also revealed that oral administration of Val-Glu-Leu-Tyr-Pro can decrease systolic blood pressure significantly (p<0.01). These results suggest that the Val-Glu-Leu-Tyr-Pro would be a beneficial ingredient for nutraceuticals and pharmaceuticals acting against hypertension and its related diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L0375000
Leucine, European Pharmacopoeia (EP) Reference Standard
C6H13NO2