EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Polyelectrolyte complexes of poly[(2-dimethylamino) ethyl methacrylate]/chondroitin sulfate obtained at different pHs: I. Preparation, characterization, cytotoxicity and controlled release of chondroitin sulfate.
PMID 25311176
Abstract
For the first time, polyelectrolyte complex based on poly[(2-dimethylamino) ethyl methacrylate] (PDMAEMA) and chondroitin sulfate (CS) was prepared. The properties of novel material and precursors were investigated by WAXS, FTIR, TGA, SEM and DLS analysis. The PDMAEMA/CS PECs presented hydrophilic-hydrophobic transition at pHs 6.0, 7.0 and 8.0 whereas the non-complexed PDMAEMA showed such a transition at pH 8.0 and not at pHs 6.0 and 7.0. Studies of CS release from PECs at pHs 6 and 8 confirmed that the samples possess the potential to release the CS in alkaline and not in acidic conditions. Since PECs are thermo-responsive due to the reduction of LCST caused by the increase in pH, the release of CS was dependent on temperature and pH factors. Cytotoxicity assays using healthy VERO cells showed that the complexation between CS and PDMAEMA increased the PECs' biocompatibility related to PDMAEMA. However, the biocompatibility depends on the amount of CS present in the PECs.