EMAIL THIS PAGE TO A FRIEND
Pharmaceutical development and technology
Amorphization of itraconazole by inorganic pharmaceutical excipients: comparison of excipients and processing methods.
PMID 25312240
Abstract
The aim of this study was to investigate the effects of solid carriers and processing routes on the properties of amorphous solid dispersions of itraconazole. Three solid carriers with a range of surface properties were studied, (1) a mesoporous silicate, magnesium aluminum silicate (Neusilin US2), (2) a nonporous silicate of corresponding composition (Veegum) and (3) a non-silicate, inorganic excipient, calcium phosphate dibasic anhydrous (A-TAB). The drug was incorporated via either solvent-deposition or ball milling. Both the maximum drug deposited by solvent-based method that produced an amorphous composite and the time for complete amorphization by co-milling was determined by X-ray powder diffraction (XRPD). Changes in the drug and excipients were monitored by nitrogen adsorption and wettability of the powder. The ability of the excipients to amorphize the drug and enhance its dissolution was related to the powder characteristics. Neusilin provided the fastest amorphization time in the mill and highest drug loading by solvent-deposition, compared with the other two excipients. Solvent-deposition provided greater dissolution enhancement than milling, due to the reduction in Neusilin porosity during high energy milling.This study confirms that substrates as well as the processing routes have notable influence on the drug deposition, amorphization, physical stability and drug in vitro release.