EMAIL THIS PAGE TO A FRIEND
Photo-dermatology
The piroxicam enigma.
PMID 2531378
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1544508
Piroxicam, United States Pharmacopeia (USP) Reference Standard
C15H13N3O4S