PloS one
Relationships between degree of polymerization and antioxidant activities: a study on proanthocyanidins from the leaves of a medicinal mangrove plant Ceriops tagal.
PMID 25313980
Abstract
Tannins from the leaves of a medicinal mangrove plant, Ceriops tagal, were purified and fractionated on Sephadex LH-20 columns. 13C nuclear magnetic resonance (13C-NMR), reversed/normal high performance liquid chromatography electrospray ionization mass spectrometry (HPLC-ESI MS) and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDT-TOF MS) analysis showed that the tannins were predominantly B-type procyanidins with minor A-type linkages, galloyl and glucosyl substitutions, and a degree of polymerization (DP) up to 33. Thirteen subfractions of the procyanidins were successfully obtained by a modified fractionation method, and their antioxidant activities were investigated using 2,2-diphenyl-1-picrylhydrazyl (DPPH) scavenging capacity and ferric reducing antioxidant power (FRAP) method. All these subfractions exhibited potent antioxidant activities, and eleven of them showed significantly different mean DP (mDP) ranging from 1.43±0.04 to 31.77±1.15. Regression analysis demonstrated that antioxidant activities were positively correlative with mDP when around mDP <10, while dropped and then remained at a level similar to mDP = 5 with around 95 µg ml(-1) for DPPH scavenging activity and 4 mmol AAE g(-1) for FRAP value.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93285
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0%
C18H12N6
T1253
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
C18H12N6
13540
Benzyl mercaptan, purum, ≥99.0% (GC)
C7H8S
B25401
Benzyl mercaptan, 99%
C7H8S
08900
Benzyl mercaptan, analytical standard
C7H8S
W214701
Benzyl mercaptan, 99%, FG
C7H8S
746487
Cesium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99.9%
ClCs
203025
Cesium chloride, ≥99.999% trace metals basis
ClCs
449733
Cesium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClCs
289329
Cesium chloride, ReagentPlus®, 99.9%
ClCs
C3139
Cesium chloride, optical grade, ≥99.5% trace metals basis
ClCs
C3011
Cesium chloride, Grade I, ≥99.0%
ClCs
562599
Cesium chloride, 99.99% trace metals basis
ClCs
562602
Cesium chloride, Cabot high-purity grade
ClCs
C3032
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
ClCs
C4036
Cesium chloride, for molecular biology, ≥98%
ClCs
C3309
Cesium chloride, BioXtra, ≥99.5% (titration)
ClCs
C6914
Cesium chloride, Grade II, ≥98%
ClCs
20966
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClCs
20967
Cesium chloride, BioUltra, ≥99.0% (AT)
ClCs
31807
Cesium chloride, puriss. p.a., ≥99.5%
ClCs