EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Enhanced spectrophotometric determination of Losartan potassium based on its physicochemical interaction with cationic surfactant.
PMID 25315864
Abstract
In this study, a simple and sensitive spectrophotometric method was developed for determination of Losartan potassium (LST K), an angiotensin-II receptor (type AT1) antagonist, in presence of cationic surfactant cetyltrimethylammonium bromide (CTAB). The physicochemical interaction of LST K with CTAB was investigated. The effect of cationic micelles on the spectroscopic and acid-base properties of LST K was studied at pH 7.4. The binding constant (Kb) and the partition coefficient (Kx) of LST K-CTAB were 1.62×10(5) M(-1) and 1.38×10(5); respectively. The binding of LST K to CTAB micelles implied a shift in drug acidity constant (ΔpKa=0.422). The developed method is linear over the range 0.5-28 μg mL(-1). The accuracy was evaluated and was found to be 99.79±0.509% and the relative standard deviation for intraday and interday precision was 0.821 and 0.963; respectively. The method was successfully applied to determine LST K in pharmaceutical formulations.