EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Effects of pentylenetetrazole kindling on mitogen-activated protein kinases levels in neocortex and hippocampus of mice.
PMID 25316496
Abstract
The epileptogenesis process involves cell signaling events associated with neuroplasticity. The mitogen-activated protein kinases (MAPKs) integrate signals originating from a variety of extracellular stimuli and may regulate cell differentiation, survival, cell death and synaptic plasticity. Here we compared the total and phosphorylated MAPKs (ERK1/2, JNK1/2 and p38(MAPK)) levels in the neocortex and hippocampus of adult Swiss male mice quantified by western blotting analysis 48xa0h after the last injection of pentylenetetrazole (PTZ), according to the kindling protocol (35xa0mg/kg, i.p., on alternated days, with a total of eight injections). The total levels of the investigated MAPKs and the phospho-p38(MAPK) in the neocortex and hippocampus were not affected by the PTZ injections. The MAPKs phosphorylation levels remain unaltered in PTZ-treated animals without convulsive seizures. The phospho-JNK2 phosphorylation, but not the phospho-JNK1, was increased in the hippocampus of PTZ-treated animals showing 1-3xa0days with convulsive seizures, whereas no significant changes were observed in those animals with more than 3xa0days with convulsive seizures. The phospho-ERK1/2 phosphorylation decreased in the neocortex and increased in the hippocampus of animals with 1-4xa0days with convulsive seizures and became unaltered in mice that showed convulsive seizures for more than 4xa0days. These findings indicate that resistance to PTZ kindling is associated with unaltered ERK1/2, JNK1/2 and p38(MAPK) phosphorylation levels in the neocortex and hippocampus. Moreover, when the PTZ kindling-induced epileptogenesis manifests behaviorally, the activation of the different MAPKs sub-families shows a variable and non-linear pattern in the neocortex and hippocampus.