EMAIL THIS PAGE TO A FRIEND
Journal of the American Society for Mass Spectrometry
Time-resolved pulsed hydrogen/deuterium exchange mass spectrometry probes gaseous proteins structural kinetics.
PMID 25318698
Abstract
A pulsed hydrogen/deuterium exchange (HDX) method has been developed for rapid monitoring of the exchange kinetics of protein ions with D2O a few milliseconds after electrospray ionization (ESI). The stepwise gradual evolution of HDX of multiply charged protein ions was monitored using the pulsed HDX mass spectrometry technique. Upon introducing a very short pulse of D2O (in the μs to ms time scale) into the linear ion trap (LIT) of a time-of-flight (TOF) mass spectrometer, bimodal distributions were detected for the ions of cytochrome c and ubiquitin. Mechanistic details of HDX reactions for ubiquitin and cytochrome c in the gas phase were uncovered and the structural transitions were followed by analyzing the kinetics of HDX.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL, analytical standard
C19H30O2
71251
Acetic acid, analytical standard
C2H4O2
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
429384
Cesium iodide, anhydrous, beads, −10 mesh, 99.999% trace metals basis
CsI
21004
Cesium iodide, analytical standard for high-resolution mass spectroscopy
CsI
203033
Cesium iodide, 99.999% trace metals basis
CsI
202134
Cesium iodide, 99.9% trace metals basis
CsI
633178
Deuterium oxide, 99.994 atom % D, contains 1 mM terephthalic acid disodium salt, 0.01 % (w/v) DSS-d6
D2O
613398
Deuterium oxide, extra, 99.994 atom % D
D2O
191701
Deuterium oxide, "100%", 99.990 atom % D
D2O
364312
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
D2O
151890
Deuterium oxide, "100%", ≥99.96 atom % D
D2O
269786
Deuterium oxide, "100%", 99.96 atom % D
D2O
151882
Deuterium oxide, 99.9 atom % D
D2O
530867
Deuterium oxide, 99.9 atom % D
D2O
613444
Deuterium oxide, 99.9 atom % D, glass distilled
D2O
450510
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
D2O
293040
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
D2O
343773
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
D2O
347167
Deuterium oxide, 99.9 atom % D, ~150 dpm/mL tritium
D2O
617385
Deuterium oxide, 99.8 atom % D
D2O
435767
Deuterium oxide, 99 atom % D
D2O
613436
Deuterium oxide, 60 atom % D
D2O
613428
Deuterium oxide, 70 atom % D
D2O
661643
Deuterium oxide, 99.99 atom % D (for D2O), contains 1% DSS-d6
D2O
756822
Deuterium oxide, filtered, 99.8 atom % D
D2O
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2