EMAIL THIS PAGE TO A FRIEND
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Protection of salvianolate against atherosclerosis via regulating the inflammation in rats.
PMID 25318872
Abstract
Inflammation plays an essential role in the pathophysiology of atherosclerosis. Our study was aimed to investigate whether salvianolate, a novel water-soluble phenolic compound of Danshen, alleviates atherosclerosis via regulating the inflammation in rats. High fat diet feeding plus vitamin D3 injection was used to induce atherosclerosis in rats. Salvianolate (60, 120 or 240 mg/kg) or placebo was given to atherosclerotic rats. The plasma lipids, interleukin 6 (IL-6) and C reactive protein (CRP) were measured by ELISA. CD4+CD25+Foxp3+ cells were determined by flow cytometry. Histological changes were examined by hematoxylin and eosin staining. The results showed that the levels of plasma IL-6 and CRP were elevated in the rats fed on high fat diet, and the histological analysis demonstrated the successful establishment of atherosclerosis models. Treatment with salvianolate alleviated the atherosclerotic process and decreased the levels of plasma IL-6 and CRP. Also the number of CD4+CD25+Foxp3+ cells was increased in salvianolate-treated rats. It was concluded that salvianolate could treat atherosclerosis via modulating the inflammation at cytokine and cell levels.