EMAIL THIS PAGE TO A FRIEND
AAPS PharmSciTech
Bilayer matrix tablets for prolonged actions of metformin hydrochloride and repaglinide.
PMID 25319054
Abstract
A combination therapy of metformin hydrochloride (MH) and repaglinide (RG) achieves a perfect glycemic control; however, the combination formulation of immediate release must be taken several times a day, compromising the therapeutic benefits and causing inconveniences to the patients. Herein, a bilayer matrix tablet that aimed at continuously releasing both MH and RG over time was developed, in which the two drugs were formulated into two separated layers. The tablets were prepared by wet granulation method, and the optimized formulation was obtained by evaluating the factors that affected the drug release. The bilayer tablets simultaneously released the two drugs over 12 h; and a better in vivo performance with a steady plasma concentration, markedly lower Cmax, prolonged Tmax, and perfect absorption was obtained. Summarily, the bilayer matrix tablets sustained both MH and RG release over time, thereby prolonging the actions for diabetic therapy and producing better health outcomes.