EMAIL THIS PAGE TO A FRIEND
Cell death & disease
TRAF1 is a key mediator for hepatic ischemia/reperfusion injury.
PMID 25321474
Abstract
Tumor necrosis factor receptor-associated factor 1 (TRAF1), an adapter in signal transduction, is involved in immunity and in apoptotic processes in various cell types. However, little is known about its function and the molecular mechanism of its activation during liver injury. This study tested the hypothesis that TRAF1 is a mediator of cell injury after hepatic ischemia/reperfusion injury (I/R). In a mouse hepatic I/R injury model, we found that TRAF1 expression was highly induced. TRAF1 deficiency was liver protective, whereas sustained TRAF1 overexpression aggravated liver injury in response to hepatic I/R injury. Mechanistic studies demonstrated that a deficiency of TRAF1 in cultured hepatocytes led to the inhibition of NF-κB-mediated inflammatory responses, suppression of the ASK/JNK pro-death pathway and promotion of cellular regeneration capacity. In contrast, the converse occurred in hepatocyte-specific TRAF1 transgenic mice. TRAF1 activated the ASK1/JNK pathway and promoted hepatic injury. Our study demonstrates that TRAF1 is a crucial early mediator of hepatic I/R injury and suggests that TRAF1 may be a potential gene therapy target for the treatment of liver injury.
Related Materials