EMAIL THIS PAGE TO A FRIEND
Bioanalysis
Application of direct-infusion ESI-MS/MS for toxicological screening.
PMID 25322781
Abstract
Direct-infusion ESI-MS/MS is a powerful approach for the identification of substances in complex mixtures. The aim of this work was to investigate its applicability to the toxicological screening of blood samples. A simple protein precipitation was used, followed by a 15 min infusion of the extract in the mass spectrometer. The application of the procedure to commercial quality controls and authentic post-mortem blood samples demonstrated that the direct-infusion ESI-MS/MS approach enables the simultaneous identification of substances that require different chromatographic conditions. However, poor sensitivity was observed for benzodiazepine, amphetamines and opiate compounds. Considering the facile implementation and positive performance of direct-infusion ESI-MS/MS, this approach may to be a valuable complementary technique for systematic toxicological analysis procedures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4144
Bromazepam, analytical standard
C14H10BrN3O
B1143000
Bromazepam, European Pharmacopoeia (EP) Reference Standard
C14H10BrN3O
C6305
Clozapine
C18H19ClN4
C171
Clozapine, solid
C18H19ClN4
C2460000
Clozapine, European Pharmacopoeia (EP) Reference Standard
C18H19ClN4
1142107
Clozapine, United States Pharmacopeia (USP) Reference Standard
C18H19ClN4
Y0000758
Clozapine for peak identification, European Pharmacopoeia (EP) Reference Standard
C18H19ClN4
1142108
Clozapine Resolution Mixture, United States Pharmacopeia (USP) Reference Standard
C18H19ClN4
C0932
Cocaethylene, analytical standard
C18H23NO4
C-010
Cocaethylene solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material
C18H23NO4
C5901
Codeine monohydrate, analytical standard
C18H21NO3 · H2O
C-006
Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material
C18H21NO3
C-015
Codeine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
C18H21NO3
C1653
Codeine solution, analytical standard, for drug analysis, 1.0 mg/mL in methanol
C18H21NO3
1303002
Haloperidol, United States Pharmacopeia (USP) Reference Standard
C21H23ClFNO2
H0100000
Haloperidol, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001518
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001517
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Z103
Zolpidem, ≥98% (HPLC), solid
C19H21N3O