EMAIL THIS PAGE TO A FRIEND
Current microbiology
Lmo0171, a novel internalin-like protein, determines cell morphology of Listeria monocytogenes and its ability to invade human cell lines.
PMID 25323012
Abstract
Internalins comprise a class of Listeria monocytogenes proteins responsible for activation of signalling pathways leading to phagocytic uptake of the bacterium by the host cell. In this paper, a possible role of Lmo0171-a new member of the internalin family was investigated. Disruption of the lmo0171 gene resulted in important cell morphology alterations along with a decrease in the ability to invade three eukaryotic cell lines, that is Int407, Hep-2 and HeLa and diminished adhesion efficiency to int407, thereby suggesting bifunctionality of the newly characterised Lmo0171 internalin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86030501
Hep2 (HeLa derivative) Cell Line human