EMAIL THIS PAGE TO A FRIEND
Nutrition research and practice
Ameliorative effect of myricetin on insulin resistance in mice fed a high-fat, high-sucrose diet.
PMID 25324935
Abstract
Obesity-associated insulin resistance is a strong risk factor for type 2 diabetes mellitus. The aim of this study was to investigate the effect of myricetin on adiposity, insulin resistance, and inflammatory markers in mice with diet-induced insulin resistance. Five-week-old male C57BL/6J mice were fed a basal diet, a high-fat, high-sucrose (HFHS) diet, or the HFHS diet containing 0.06% myricetin or 0.12% myricetin for 12 weeks after a 1-week adaptation, and body weight and food intake were monitored. After sacrifice, serum lipid profiles, glucose, insulin, adipocyte-derived hormones, and proinflammatory cytokines were measured. The homeostasis model assessment for insulin resistance (HOMA-IR) was determined. Myricetin given at 0.12% of the total diet significantly reduced body weight, weight gain, and epidydimal white adipose tissue weight, and improved hypertriglyceridemia and hypercholesterolemia without a significant influence on food intake in mice fed the HFHS diet. Serum glucose and insulin levels, as well as HOMA-IR values, decreased significantly by 0.12% myricetin supplementation in mice fed the HFHS diet. Myricetin given at 0.12% of the total diet significantly reduced serum levels of leptin, tumor necrosis factor-α (TNF-α) and interleukin-6 (IL-6) in mice fed the HFHS diet. These findings suggest that myricetin may have a protective effect against diet-induced obesity and insulin resistance in mice fed HFHS diet, and that alleviation of insulin resistance could partly occur by improving obesity and reducing serum proinflammatory cytokine levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72576
Myricetin, analytical standard
C15H10O8
112941
tert-Butylhydroquinone, 97%
C10H14O2