EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Inhibition of human aldehyde oxidase activity by diet-derived constituents: structural influence, enzyme-ligand interactions, and clinical relevance.
PMID 25326286
Abstract
The mechanistic understanding of interactions between diet-derived substances and conventional medications in humans is nascent. Most investigations have examined cytochrome P450-mediated interactions. Interactions mediated by other phase I enzymes are understudied. Aldehyde oxidase (AO) is a phase I hydroxylase that is gaining recognition in drug design and development programs. Taken together, a panel of structurally diverse phytoconstituents (n = 24) was screened for inhibitors of the AO-mediated oxidation of the probe substrate O(6)-benzylguanine. Based on the estimated IC50 (<100 μM), 17 constituents were advanced for Ki determination. Three constituents were described best by a competitive inhibition model, whereas 14 constituents were described best by a mixed-mode model. The latter model consists of two Ki terms, Kis and Kii, which ranged from 0.26-73 and 0.80-120 μM, respectively. Molecular modeling was used to glean mechanistic insight into AO inhibition. Docking studies indicated that the tested constituents bound within the AO active site and elucidated key enzyme-inhibitor interactions. Quantitative structure-activity relationship modeling identified three structural descriptors that correlated with inhibition potency (r(2) = 0.85), providing a framework for developing in silico models to predict the AO inhibitory activity of a xenobiotic based solely on chemical structure. Finally, a simple static model was used to assess potential clinically relevant AO-mediated dietary substance-drug interactions. Epicatechin gallate and epigallocatechin gallate, prominent constituents in green tea, were predicted to have moderate to high risk. Further characterization of this uncharted type of interaction is warranted, including dynamic modeling and, potentially, clinical evaluation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52186
(±)-Naringenin, analytical standard
C15H12O5
50299
(−)-Epigallocatechin gallate, ≥97.0% (HPLC)
C22H18O11
93894
(−)-Epigallocatechin gallate, analytical standard
C22H18O11
M1381
4-Methylumbelliferone, ≥98%
C10H8O3
42251
Apigenin Proteus mirabilis, analytical standard
C15H10O5
01760595
Apigenin Proteus mirabilis, primary pharmaceutical reference standard
C15H10O5
03970590
Epigallocatechin gallate, primary pharmaceutical reference standard
C22H18O11
PHR1333
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
C22H18O11
H5254
Hesperidin, ≥80%
C28H34O15
50162
Hesperidin, analytical standard
C28H34O15
Y0001203
Hesperidin, European Pharmacopoeia (EP) Reference Standard
C28H34O15
04650590
Hesperidin, primary pharmaceutical reference standard
C28H34O15
Y0000011
Hymecromone
C10H8O3
96353
Kaempferol, analytical standard
C15H10O6
00550580
Kaempferol, primary pharmaceutical reference standard
C15H10O6
N1376
Naringin, ≥90%, from citrus fruit
C27H32O14
71162
Naringin, ≥95% (HPLC)
C27H32O14
91842
Naringin, analytical reference material
C27H32O14
Y0001378
Naringin, European Pharmacopoeia (EP) Reference Standard
C27H32O14
P8399
Psoralen, ≥99% (HPLC)
C11H6O3
73633
Psoralen, analytical standard
C11H6O3
T0891
Tolbutamide, analytical standard
C12H18N2O3S
46968
Tolbutamide, VETRANAL, analytical standard
C12H18N2O3S
T1700000
Tolbutamide, European Pharmacopoeia (EP) Reference Standard
C12H18N2O3S