EMAIL THIS PAGE TO A FRIEND
Nanoscale research letters
Facile synthesis of folate-conjugated magnetic/fluorescent bifunctional microspheres.
PMID 25328504
Abstract
In this paper, we investigated the functional imaging properties of magnetic microspheres composed of magnetic core and CdTe quantum dots in the silica shell functionalized with folic acid (FA). The preparation procedure included the preparation of chitosan-coated Fe3O4 nanoparticles (CS-coated Fe3O4 NPs) prepared by a one-pot solvothermal method, the reaction between carboxylic and amino groups under activation of NHS and EDC in order to obtain the CdTe-CS-coated Fe3O4 NPs, and finally the growth of SiO2 shell vent the photoluminescence (PL) quenching via a Stöber method (Fe3O4-CdTe@SiO2). Moreover, in order to have a specific targeting capacity, the magnetic and fluorescent bifunctional microspheres were synthesized by bonding of SiO2 shell with FA molecules via amide reaction (Fe3O4-CdTe@SiO2-FA). The morphology, size, chemical components, and magnetic property of as-prepared composite nanoparticles were characterized by ultraviolet-visible spectroscopy, fluorescent spectroscopy, Fourier transform infrared spectroscopy (FTIR), X-ray powder diffraction (XRD), scanning transmission electron microscopy (SEM), transmission electron microscopy (TEM), thermogravimetric analysis (TGA), and vibrating sample magnetometer (VSM), respectively. The results show that the magnetic and fluorescent bifunctional microspheres have strong luminescent which will be employed for immuno-labeling and fluorescent imaging of HeLa cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63768
3-Mercaptopropionic acid, ≥99.0% (HPLC)
C3H6O2S
M5801
3-Mercaptopropionic acid, ≥99%
C3H6O2S
439800
Cadmium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
CdCl2
202908
Cadmium chloride, 99.99% trace metals basis
CdCl2
655198
Cadmium chloride, technical grade
CdCl2
324558
Ethylene glycol, anhydrous, 99.8%
C2H6O2
03747
Ethylene glycol, BioUltra, ≥99.5% (GC)
C2H6O2
85978
Ethylene glycol, analytical standard
C2H6O2
1265515
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
C2H6O2
PHR1046
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O2
24204
Ethylene glycol, puriss., ≥99.5% (GC)
C2H6O2
61941 Ethylene glycol 5 M solution
C2H6O2
721980 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
C2H6O2
721972 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
C2H6O2
236489
Iron(III) chloride hexahydrate, ACS reagent, 97%
Cl3Fe · 6H2O
F2877
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Cl3Fe · 6H2O
44944
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Cl3Fe · 6H2O
31232
Iron(III) chloride hexahydrate, puriss. p.a., Reag. Ph. Eur., ≥99%
Cl3Fe · 6H2O
56480
N-Hydroxysuccinimide, purum, ≥97.0% (T)
C4H5NO3
791741
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3NaO2
S2889
Sodium acetate, anhydrous, for molecular biology, ≥99%
C2H3NaO2
S3272
Sodium acetate, for electrophoresis, ≥99%, crystalline
C2H3NaO2
S5636
Sodium acetate, powder, BioReagent, for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
C2H3NaO2
S1429
Sodium acetate, meets USP testing specifications, anhydrous
C2H3NaO2
1613407
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
C2H3NaO2
229873
Sodium acetate, 99.995% trace metals basis
C2H3NaO2
S7545
Sodium acetate, BioXtra, ≥99.0%
C2H3NaO2
110191
Sodium acetate, ReagentPlus®, 99%
C2H3NaO2
71183
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
C2H3NaO2
W302406
Sodium Acetate Anhydrous, >99%, FCC, FG
C2H3NaO2