EMAIL THIS PAGE TO A FRIEND
Pharmaceutical biology
Chemical profile and biological activities of Veronica thymoides subsp. pseudocinerea.
PMID 25331745
Abstract
In Turkey, Veronica species (Plantaginaceae) have been used as a diuretic and for wound healing in traditional medicine. To examine the fatty acid and essential oil profiles, the antioxidant, anticholinesterase, antimicrobial, and DNA damage effects of Veronica thymoides P.H. Davis subsp. pseudocinerea M.A. Fischer as a potential source of natural active compounds. GC/MS was used to analyze essential oil and fatty acid obtained from whole plant. The antioxidant activity was evaluated by the β-carotene-linoleic acid test system, DPPH-free and ABTS cation radicals scavenging, and cupric reducing antioxidant capacity assays. The anticholinesterase and antimicrobial activities were determined by Ellman and broth macrodillution methods, respectively. The effect of the methanol extract on DNA cleavage was investigated. Hexatriacontene (21.0%) was found to be the main constituent in essential oil, and linoleic acid (25.2%) and palmitic acid (20.6%) in fatty acid. Methanol extract demonstrated the best IC50 values in lipid peroxidation (49.81 ± 0.31 µg/ml) and DPPH-free radical scavenging activity (15.32 ± 0.17 µg/ml). Methanol and water extracts possessed strong ABTS cation radical scavenging activity with IC50 values 9.15 ± 0.28 and 8.90 ± 0.1  µg/ml, respectively. The acetone extract exhibited moderate butyrylcholinesterase inhibitory activity. The highest antimicrobial activity was determined in methanol extract against Escherichia coli with 31.25 µg/ml MIC value. Inhibition of methanol extract on plasmid DNA cleavage by OH radicals was found to be 93.32% at 500 µg/ml. The methanol extract having strong antioxidant and DNA damage effects could be investigated phytochemically to find natural active compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
Y0001470
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O2
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
33352
Methanolic Base, 0.5 M CH3ONa (0.5N), for GC derivatization
CH3NaO
33080
Methanolic Base, 0.5 M CH3ONa (0.5N), for GC derivatization
CH3NaO
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
N1501
Neocuproine, ≥98%
C14H12N2
791733
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3KO2
60489
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
K2O8S2
B3253
S-Butyrylthiocholine iodide, ≥98%
C9H20INOS
20820
S-Butyrylthiocholine iodide, puriss., ≥99.0% (AT)
C9H20INOS
164992
Sodium methoxide, reagent grade, 95%, powder
CH3NaO
71748
Sodium methoxide solution, technical, ~30% in methanol (T, ~5.4 M)
CH3NaO
403067
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
CH3NaO
156256
Sodium methoxide solution, 25 wt. % in methanol
CH3NaO