EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
An intramolecular lock facilitates folding and stabilizes the tertiary structure of Streptococcus mutans adhesin P1.
PMID 25331888
Abstract
The cariogenic bacterium Streptococcus mutans uses adhesin P1 to adhere to tooth surfaces, extracellular matrix components, and other bacteria. A composite model of P1 based on partial crystal structures revealed an unusual complex architecture in which the protein forms an elongated hybrid alpha/polyproline type II helical stalk by folding back on itself to display a globular head at the apex and a globular C-terminal region at the base. The structure of P1's N terminus and the nature of its critical interaction with the C-terminal region remained unknown, however. We have cocrystallized a stable complex of recombinant N- and C-terminal fragments and here describe a previously unidentified topological fold in which these widely discontinuous domains are intimately associated. The structure reveals that the N terminus forms a stabilizing scaffold by wrapping behind the base of P1's elongated stalk and physically "locking" it into place. The structure is stabilized through a highly favorable ΔG(solvation) on complex formation, along with extensive hydrogen bonding. We confirm the functional relevance of this intramolecular interaction using differential scanning calorimetry and circular dichroism to show that disruption of the proper spacing of residues 989-1001 impedes folding and diminishes stability of the full-length molecule, including the stalk. Our findings clarify previously unexplained functional and antigenic properties of P1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2105686
Anti-A4GALT antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2105151
Anti-B3GALNT1 antibody produced in rabbit, affinity isolated antibody
HPA041204
Anti-B3GALNT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106690
Anti-B3GALNT1 antibody produced in rabbit, affinity isolated antibody
AV41764
Anti-GNAS (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV41765
Anti-GNAS (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV42694
Anti-GNAS (AB3) antibody produced in rabbit, IgG fraction of antiserum
SAB2501411
Anti-GNAS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA018122
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100941
Anti-GNAS antibody produced in rabbit, affinity isolated antibody
HPA027478
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028386
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
AV36644
Anti-MCM3 (AB1) antibody produced in rabbit, affinity isolated antibody
AV36645
Anti-MCM3 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1412541
ANTI-MCM3 antibody produced in mouse, clone 2A11, purified immunoglobulin, buffered aqueous solution
SAB1412669
ANTI-MCM3 antibody produced in mouse, clone 2H3, purified immunoglobulin, buffered aqueous solution
SAB2701703
Anti-mcm3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004789
Anti-MCM3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004790
Anti-MCM3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1303779
ANTI-NUCKS1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4502774
Anti-NUCKS1 antibody produced in rabbit, affinity isolated antibody
SAB1406291
Anti-PRF1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV42208
Anti-PRF1 antibody produced in rabbit, affinity isolated antibody
SAB1401303
Anti-PRF1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406308
Anti-PRM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA055150
Anti-PRM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406403
Anti-RPLP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003368
Anti-RPLP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406441
Anti-SILV antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV46802
Anti-SILV antibody produced in rabbit, IgG fraction of antiserum