EMAIL THIS PAGE TO A FRIEND
Kidney international
Insulin resistance in chronic kidney disease is ameliorated by spironolactone in rats and humans.
PMID 25337775
Abstract
In this study, we examined the association between chronic kidney disease (CKD) and insulin resistance. In a patient cohort with nondiabetic stages 2-5 CKD, estimated glomerular filtration rate (eGFR) was negatively correlated and the plasma aldosterone concentration was independently associated with the homeostasis model assessment of insulin resistance. Treatment with the mineralocorticoid receptor blocker spironolactone ameliorated insulin resistance in patients, and impaired glucose tolerance was partially reversed in fifth/sixth nephrectomized rats. In these rats, insulin-induced signal transduction was attenuated, especially in the adipose tissue. In the adipose tissue of nephrectomized rats, nuclear mineralocorticoid receptor expression, expression of the mineralocorticoid receptor target molecule SGK-1, tissue aldosterone content, and expression of the aldosterone-producing enzyme CYP11B2 increased. Mineralocorticoid receptor activation in the adipose tissue was reversed by spironolactone. In the adipose tissue of nephrectomized rats, asymmetric dimethylarginine (ADMA; an uremic substance linking uremia and insulin resistance) increased, the expression of the ADMA-degrading enzymes DDAH1 and DDAH2 decreased, and the oxidative stress increased. All of these changes were reversed by spironolactone. In mature adipocytes, aldosterone downregulated both DDAH1 and DDAH2 expression, and ADMA inhibited the insulin-induced cellular signaling. Thus, activation of mineralocorticoid receptor and resultant ADMA accumulation in adipose tissue has, in part, a relevant role in the development of insulin resistance in CKD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4268
NG,NG-Dimethylarginine dihydrochloride
C8H18N4O2 · 2HCl