PloS one
Presynaptic localization of Smn and hnRNP R in axon terminals of embryonic and postnatal mouse motoneurons.
PMID 25338097
Abstract
Spinal muscular atrophy (SMA) is caused by deficiency of the ubiquitously expressed survival motoneuron (SMN) protein. SMN is crucial component of a complex for the assembly of spliceosomal small nuclear ribonucleoprotein (snRNP) particles. Other cellular functions of SMN are less characterized so far. SMA predominantly affects lower motoneurons, but the cellular basis for this relative specificity is still unknown. In contrast to nonneuronal cells where the protein is mainly localized in perinuclear regions and the nucleus, Smn is also present in dendrites, axons and axonal growth cones of isolated motoneurons in vitro. However, this distribution has not been shown in vivo and it is not clear whether Smn and hnRNP R are also present in presynaptic axon terminals of motoneurons in postnatal mice. Smn also associates with components not included in the classical SMN complex like RNA-binding proteins FUS, TDP43, HuD and hnRNP R which are involved in RNA processing, subcellular localization and translation. We show here that Smn and hnRNP R are present in presynaptic compartments at neuromuscular endplates of embryonic and postnatal mice. Smn and hnRNP R are localized in close proximity to each other in axons and axon terminals both in vitro and in vivo. We also provide new evidence for a direct interaction of Smn and hnRNP R in vitro and in vivo, particularly in the cytosol of motoneurons. These data point to functions of SMN beyond snRNP assembly which could be crucial for recruitment and transport of RNA particles into axons and axon terminals, a mechanism which may contribute to SMA pathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB144P
Anti-Choline Acetyltransferase Antibody, Anti-Choline Acetyltransferase Antibody detects level of ChAT and has been published and validated for use in IH(P), IC, IH and WB.
AB5539
Anti-Neurofilament H Antibody, Anti-Neurofilament H Antibody is an antibody against Neurofilament for use in IC, IH, IH(P) & WB.
SAB2700924
Anti-HNRNPR (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA026092
Anti-HNRNPR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101057
Anti-HNRNPR antibody produced in rabbit, affinity isolated antibody
B3773 Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
CKOZFN10210 CompoZr® Knockout ZFN Kit, Human HNRNPR (NM_001102397)
CKOZFND10210 CompoZr® Knockout ZFN Kit, ZFN plasmid only, Human HNRNPR (NM_001102397)
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
G2904 Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
A8062 Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148 Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891 Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
A9452 Monoclonal Anti-Goat/Sheep IgG–Peroxidase antibody produced in mouse, clone GT-34, purified immunoglobulin, lyophilized powder
I6135 Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
I6260 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
I6760 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
F5016 Monoclonal Anti-Human IgG (Fc specific)−FITC antibody produced in mouse, clone HP-6017, purified immunoglobulin, buffered aqueous solution
I5885 Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
I9513 Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
I9888 Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
R0761 Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid