PloS one
Dextromethorphan mediated bitter taste receptor activation in the pulmonary circuit causes vasoconstriction.
PMID 25340739
Abstract
Activation of bitter taste receptors (T2Rs) in human airway smooth muscle cells leads to muscle relaxation and bronchodilation. This finding led to our hypothesis that T2Rs are expressed in human pulmonary artery smooth muscle cells and might be involved in regulating the vascular tone. RT-PCR was performed to reveal the expression of T2Rs in human pulmonary artery smooth muscle cells. Of the 25 T2Rs, 21 were expressed in these cells. Functional characterization was done by calcium imaging after stimulating the cells with different bitter agonists. Increased calcium responses were observed with most of the agonists, the largest increase seen for dextromethorphan. Previously in site-directed mutational studies, we have characterized the response of T2R1 to dextromethorphan, therefore, T2R1 was selected for further analysis in this study. Knockdown with T2R1 specific shRNA decreased mRNA levels, protein levels and dextromethorphan-induced calcium responses in pulmonary artery smooth muscle cells by up to 50%. To analyze if T2Rs are involved in regulating the pulmonary vascular tone, ex vivo studies using pulmonary arterial and airway rings were pursued. Myographic studies using porcine pulmonary arterial and airway rings showed that stimulation with dextromethorphan led to contraction of the pulmonary arterial and relaxation of the airway rings. This study shows that dextromethorphan, acting through T2R1, causes vasoconstrictor responses in the pulmonary circuit and relaxation in the airways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3755
6-Propyl-2-thiouracil, enzyme inhibitor
C7H10N2OS
46698
6-Propyl-2-thiouracil, VETRANAL, analytical standard
C7H10N2OS
82460
6-Propyl-2-thiouracil, purum, ≥98.0% (T)
C7H10N2OS
C2580000
Colchicine, (European Pharmacopoeia (EP) Reference Standard)
C22H25NO6
S0625
D-(−)-Salicin, ≥99% (GC)
C13H18O7
79588
D-(−)-Salicin, analytical standard
C13H18O7
D5765
Denatonium benzoate, ≥98%
C28H34N2O3
1180503 Dextromethorphan, United States Pharmacopeia (USP) Reference Standard
C18H25NO
EHU142701 MISSION® esiRNA, esiRNA human TAS2R1 (esiRNA1)
P7629
N-Phenylthiourea, ≥98%
C7H8N2S
P5272
N-Phenylthiourea, Grade II, ≥90% (HPLC)
C7H8N2S
31056
N-Phenylthiourea, PESTANAL®, analytical standard
C7H8N2S
222909
N-Phenylthiourea, 97%
C7H8N2S
22620
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
C20H24N2O2
145904
Quinine, 90%
C20H24N2O2
69311
Quinine, certified reference material, TraceCERT®
C20H24N2O2
Y0000669
Salicin, European Pharmacopoeia (EP) Reference Standard
C13H18O7
467871
Sodium thiocyanate, ≥99.99% trace metals basis
CNNaS
251410
Sodium thiocyanate, ACS reagent, ≥98.0%
CNNaS
S7757
Sodium thiocyanate, reagent grade, 98-102% (titration)
CNNaS
08536
Sodium thiocyanate 2 M solution
CNNaS
80518
Sodium thiocyanate solution, BioUltra, 8 M in H2O
CNNaS