EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Forsythiaside protects against hydrogen peroxide-induced oxidative stress and apoptosis in PC12 cell.
PMID 25344274
Abstract
Oxidative stress is a major component of harmful cascades activated in neurodegenerative disorders. We sought to elucidate possible effects of forsythiaside, an active constituent isolated from the Chinese medicinal herb Forsythia suspense, on hydrogen peroxide (H2O2)-induced cell death and to determine the underlying molecular mechanisms in neuron-like PC12 cells. We found that forsythiaside treatment effectively protected PC12 cells against H2O2-induced cell damage and apoptosis. H2O2 exposure induced oxidative stress in PC12 cells, as revealed by increased ROS and lipid peroxidation (MDA), which were inhibited by forsythiaside pretreatment. Increased Bax/Bcl-2 ratio, mitochondrial membrane potential decrease, cytochrome c release, caspase-9/-3 activation, AIF/Endo G translocation were observed in H2O2-treated cells. Interestingly, forsythiaside effectively prevented these events. These results suggested that forsythiaside prevented H2O2-induced mitochondria-dependent apoptosis. Further, increased nuclear levels of Nrf2 and up-regulation of antioxidant enzymes (Mn/SOD and CAT) were detected in forsythiaside-treated cells, indicating the anti-oxidative effects of forsythiaside might be associated with activation of Nrf2 pathway. Moreover, forsythiaside was proved to be effective to prevent LPS-induced cell death and ROS generation. In conclusion, forsythiaside effectively inhibited H2O2-induced oxidative stress and subsequent apoptosis activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31640
4,4′-Diaminodiphenylmethane, analytical standard
C13H14N2