EMAIL THIS PAGE TO A FRIEND
Annals of oncology : official journal of the European Society for Medical Oncology
A phase IB trial of the oral MEK inhibitor trametinib (GSK1120212) in combination with everolimus in patients with advanced solid tumors.
PMID 25344362
Abstract
This phase Ib trial investigated the safety, tolerability, and recommended phase II dose and schedule of the MEK inhibitor trametinib in combination with the mammalian target of rapamycin (mTOR) inhibitor everolimus. Secondary objectives included pharmacokinetic (PK) characterization and evaluation of clinical activity. A total of 67 patients with advanced solid tumors were enrolled in this open-label, single-arm, dose-escalation study. Dose escalation followed a 3 + 3 design. Patients were assigned to one of 10 different cohorts, involving either daily dosing with both agents or daily dosing with trametinib and intermittent everolimus dosing. This included an expansion cohort comprising patients with pancreatic tumors. PKs samples were collected predose, as well as 1, 2, 4, and 6 h post-dose on day 15 of the first treatment cycle. Concurrent treatment with trametinib and everolimus resulted in frequent treatment-related adverse events, including mucosal inflammation (40%), stomatitis (25%), fatigue (54%), and diarrhea (42%). PK assessment did not suggest drug-drug interactions between these two agents. Of the 67 enrolled patients, 5 (7%) achieved partial response (PR) to treatment and 21 (31%) displayed stable disease (SD). Among the 21 patients with pancreatic cancer, PR was observed in 1 patient (5%) and SD in 6 patients (29%). This study was unable to identify a recommended phase II dose and schedule of trametinib in combination with everolimus that provided an acceptable tolerability and adequate drug exposure.