EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
Thymoquinone Pretreatment Overcomes the Insensitivity and Potentiates the Antitumor Effect of Gemcitabine Through Abrogation of Notch1, PI3K/Akt/mTOR Regulated Signaling Pathways in Pancreatic Cancer.
PMID 25344906
Abstract
The gemcitabine-insensitivity remains the main challenge for pancreatic cancer treatment. Thymoquinone, the predominant bioactive ingredient of Nigella sativa, has been shown to possess promising anti-cancer and chemo-sensitizing effects on pancreatic cancer, however, its meticulous mechanism is still indistinct. The objective of the present study was to investigate the potency of thymoquinone in combination with gemcitabine in inducing apoptosis and preventing the development of gemcitabine-insensitivity in pancreatic cancer cells. The anti-tumor effects of thymoquinone and gemcitabine were analyzed via evaluation of alterations of cell viability, tumor weight, apoptosis-related proteins, caspase-3, -9 activities and NF-κB DNA binding activity in pancreatic cancer cells in vitro and PANC-1 cells orthotopic xenograft in vivo. Thymoquinone pretreatment following gemcitabine treatment synergistically caused an increase in pancreatic cancer cells apoptosis and tumor growth inhibition both in vitro and in vivo. The novel combinational regimen also contributes to alterations of multiple molecular signaling targets, such as the suppression of Notch1, NICD accompanying with up-regulation of PTEN, the inactivation of Akt/mTOR/S6 signaling pathways, and the suppression of phosphorylation and nuclear translocation of p65 induced by TNF-α. Thymoquinone pretreatment and gemcitabine also induced down-regulation of anti-apoptotic Bcl-2, Bcl-xL, XIAP and up-regulation and activation of pro-apoptotic molecules including Caspase-3, Caspase-9, Bax and increased release of cytochrome c. This novel modality of thymoquinone pretreatment can enhance the anti-cancer activity of gemcitabine and may be a promising option in the treatment of pancreatic cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274666
Thymoquinone, ≥98%
C10H12O2