EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
Determination of ethanol using permanganate-CdS quantum dot chemiluminescence system.
PMID 25345928
Abstract
A novel and highly sensitive chemiluminescence (CL) method for the determination of ethanol was developed based on the CdS quantum dots (QDs)-permanganate system. It was found that KMnO4 could directly oxidize CdS QDs in acidic media resulting in relatively high CL emission. A possible mechanism was proposed for this reaction based on UV/Vis absorption, fluorescence and the generated CL emission spectra. However, it was observed that ethanol had a remarkable inhibition effect on this system. This effect was exploited in the determination of ethanol within the concentration range 12-300 µg/L, with detection at 4.3 µg/L. In order to evaluate the capability of presented method, it was satisfactorily utilized in the determination of alcohol in real samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
407410
Sodium sulfide
Na2S
13468
Sodium sulfide hydrate, ≥60%, scales
Na2S · xH2O