EMAIL THIS PAGE TO A FRIEND
Nanoscale
Ostwald-ripening and particle size focussing of sub-10 nm NaYF₄ upconversion nanocrystals.
PMID 25347027
Abstract
We have studied the growth behaviour of sub-10 nm NaYF₄ upconversion nanocrystals of the hexagonal β-phase and the cubic α-phase. Ostwald-ripening of such particles in oleic acid/octadecene solvent results in broadening of the particle size distribution if the colloid contains particles of one crystal phase only. Narrow size distributions are formed only if β-phase particles grow in the presence of an excess of α-phase particles. Such binary mixtures of α-phase and β-phase particles form intrinsically when colloids of α-phase particles are heated for a sufficiently long time, because seeds of the β-phase nucleate in the solution after some time at high temperatures. Since the number of seeds determines the final size of the β-phase product, control of the nucleation is crucial for controlling the final particle size. We show that the number of β-phase seeds strongly depends on the composition of the α-phase known to form solid solutions Na₁-xYF₄-x in the range from x = 0 to x = 4/9. Sodium-deficient α-phase particles form a negligible number of β-phase seeds whereas α-phase particles with high sodium content yield a very large number of seeds. By taking advantage of this dependence and modifying the synthesis of the α-phase particles accordingly, small phase-pure β-NaYF₄:Yb,Er particles with a size smaller than 6 nm can be prepared in oleic acid/octadecene just as well as much larger particles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74738
1-Octadecene, analytical standard
C18H36
74740
1-Octadecene, ≥95.0% (GC)
C18H36
O806
1-Octadecene, technical grade, 90%
C18H36
74745
1-Octadecene, technical, ~85% (GC)
C18H36
338869
Ammonium fluoride, ≥99.99% trace metals basis
H4FN
216011
Ammonium fluoride, ACS reagent, ≥98.0%
H4FN
30101
Ammonium fluoride, puriss. p.a., ACS reagent, ≥98%
H4FN
40292
Ammonium fluoride solution, semiconductor grade VLSI PURANAL (Honeywell 17600), 40%
H4FN
263052
Erbium, chips, 99.9% trace rare earth metals basis
Er
693685
Erbium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Er
261084
Erbium, ingot, 99.9% trace rare earth metals basis
Er
263044
Erbium, powder, −40 mesh, 99.9% trace rare earth metals basis
Er
755990
Erbium, dendritic pieces, purified by distillation, 99.95% trace metals basis
Er
GF45743698
Erbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Er
GF15099767
Erbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Er
GF75586494
Erbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Er
GF59778152
Erbium, foil, 50x50mm, thickness 1.0mm, as rolled, 99%
Er
GF14957994
Erbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Er
GF09631043
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Er
GF93107189
Erbium, rod, 100mm, diameter 10.0mm, cast, 99%
Er
GF34237469
Erbium, rod, 100mm, diameter 6.35mm, cast, 99%
Er
GF21606386
Erbium, rod, 50mm, diameter 10.0mm, cast, 99%
Er
GF18382248
Erbium, rod, 50mm, diameter 2.0mm, cast, 99%
Er
GF33873891
Erbium, rod, 50mm, diameter 6.35mm, cast, 99%
Er
GF87470253
Erbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Er
GF75856225
Erbium, foil, 25x25mm, thickness 1.0mm, as rolled, 99%
Er
75090
Oleic acid, analytical standard
C18H34O2
Y0001479
Oleic acid, European Pharmacopoeia (EP) Reference Standard
C18H34O2
05508
Oleic acid, Selectophore, ≥99.0%
C18H34O2
O7501
Sodium oleate, ≥99%
C18H33NaO2