EMAIL THIS PAGE TO A FRIEND
eLife
CD28 expression is required after T cell priming for helper T cell responses and protective immunity to infection.
PMID 25347065
Abstract
The co-stimulatory molecule CD28 is essential for activation of helper T cells. Despite this critical role, it is not known whether CD28 has functions in maintaining T cell responses following activation. To determine the role for CD28 after T cell priming, we generated a strain of mice where CD28 is removed from CD4(+) T cells after priming. We show that continued CD28 expression is important for effector CD4(+) T cells following infection; maintained CD28 is required for the expansion of T helper type 1 cells, and for the differentiation and maintenance of T follicular helper cells during viral infection. Persistent CD28 is also required for clearance of the bacterium Citrobacter rodentium from the gastrointestinal tract. Together, this study demonstrates that CD28 persistence is required for helper T cell polarization in response to infection, describing a novel function for CD28 that is distinct from its role in T cell priming.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9304 Anti-Bcl-x antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500349
Anti-Bcl-xL antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
GW21157 Anti-BCL2L1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV02008
Anti-BCL2L1 antibody produced in rabbit, affinity isolated antibody
AV30475
Anti-BCL2L1 antibody produced in rabbit, IgG fraction of antiserum
SAB2103893
Anti-BCL2L1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1408550
Anti-BLR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C1366 Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501241
Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody
SAB3501088
Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody
C3366 Anti-CXCR5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA042432
Anti-CXCR5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401658
Anti-ICOS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22461F Anti-Interleukin-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I8398 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I8273 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I7401 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7784 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
B9429
Monoclonal Anti-Bcl-xL antibody produced in mouse, ~2 mg/mL, clone 2H12, purified immunoglobulin, buffered aqueous solution
WH0000598M1
Monoclonal Anti-BCL2L1 antibody produced in mouse, clone 3E2-2A5, purified immunoglobulin, buffered aqueous solution
WH0000643M1
Monoclonal Anti-BLR1 antibody produced in mouse, clone 2C1, purified immunoglobulin, buffered aqueous solution
C6473 Monoclonal Anti-CXCR3 antibody produced in mouse, clone 49801.111, purified immunoglobulin, lyophilized powder
SAB1402794
Monoclonal Anti-CXCR3, (N-terminal) antibody produced in mouse, clone 1C5, ascites fluid
WH0029851M1 Monoclonal Anti-ICOS antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412835
Monoclonal Anti-ICOS antibody produced in mouse, clone 3G4, purified immunoglobulin, 0.5 mg/mL
WH0003558M3 Monoclonal Anti-IL2 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution
I6784 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5334, purified immunoglobulin, lyophilized powder
I6909 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5355, purified immunoglobulin, lyophilized powder