EMAIL THIS PAGE TO A FRIEND
Journal of neuro-oncology
PD-1, PD-L1, PD-L2 expression in the chordoma microenvironment.
PMID 25349132
Abstract
Chordomas are rare malignant tumors that are postulated to arise from remnants of the notochord. Currently, the interaction between chordomas and the host immune system is poorly understood. The checkpoint protein, PD-1 is expressed by circulating lymphocytes and is a marker of activation and exhaustion. Its ligands, PD-L1 (B7-H1, CD274) and PD-L2 (B7-DC, CD273), are expressed on a variety of human cancers; however this pathway has not been previously reported in chordomas. We used flow cytometric and RT-PCR analysis in three established primary and recurrent chordoma cell lines (U-CH1, U-CH2, and JHC7) as well as immunohistochemical analysis of chordoma tissues from 10 patients to identify and localize expression of PD-1 pathway proteins. PD-1 ligands are not constitutively expressed by chordoma cells, but their expression is induced in the setting of pro-inflammatory cytokines in all cell lines examined. In paraffin embedded tissues, we found that tumor infiltrating lymphocytes expressed PD-1 in 3/6 cases. We also found that, although chordoma cells did not express significant levels of PD-L1, PD-L1 expression was observed on tumor-infiltrating macrophages and tumor infiltrating lymphocytes. Our study suggests that PD-1, PD-L1, and PD-L2 are present in the microenvironment of a subset of chordomas analyzed. Future studies are needed to evaluate the contribution of the PD-1 pathway to the immunosuppressive microenvironment of chordomas.