EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
The advantage of antibody cocktails for targeted alpha therapy depends on specific activity.
PMID 25349219
Abstract
Nonuniform dose distributions among disseminated tumor cells can be a significant limiting factor in targeted α therapy. This study examines how cocktails of radiolabeled antibodies can be formulated to overcome this limitation. Cultured MDA-MB-231 human breast cancer cells were treated with different concentrations of a cocktail of 4 fluorochrome-conjugated monoclonal antibodies. The amount of each antibody bound to each cell was quantified using flow cytometry. A spreadsheet was developed to "arm" the antibodies with any desired radionuclide and specific activity, calculate the absorbed dose to each cell, and perform a Monte Carlo simulation of the surviving fraction of cells after exposure to cocktails of different antibody combinations. Simulations were performed for the α-particle emitters (211)At, (213)Bi, and (225)Ac. Activity delivered to the least labeled cell can be increased by 200%-400% with antibody cocktails, relative to the best-performing single antibody. Specific activity determined whether a cocktail or a single antibody achieved greater cell killing. With certain specific activities, cocktails outperformed single antibodies by a factor of up to 244. There was a profound difference (≤16 logs) in the surviving fraction when a uniform antibody distribution was assumed and compared with the experimentally observed nonuniform distribution. These findings suggest that targeted α therapy can be improved with customized radiolabeled antibody cocktails. Depending on the antibody combination and specific activity of the radiolabeled antibodies, cocktails can provide a substantial advantage in tumor cell killing. The methodology used in this analysis provides a foundation for pretreatment prediction of tumor cell survival in the context of personalized cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF86561354
Bismuth, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF85045631
Bismuth, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF80139438
Bismuth, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF81875447
Bismuth, rod, 120mm, diameter 10.0mm, cast sticks, 100%
Bi
GF71052432
Bismuth, rod, 140mm, diameter 10.0mm, cast sticks, 100%
Bi
GF60966366
Bismuth, rod, 70mm, diameter 10.0mm, cast sticks, 100%
Bi
GF89918500
Bismuth, foil, 25x25mm, thickness 0.25mm, 100%
Bi
GF71570550
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Bi
GF58555581
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Bi
GF52383506
Bismuth, foil, 25x25mm, thickness 1.0mm, 99.97%
Bi
GF31001960
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Bi
GF39813419
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Bi
GF46319340
Bismuth, foil, 50x50mm, thickness 0.5mm, 100%
Bi
GF19448017
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Bi
GF97656238
Bismuth, foil, 50x50mm, thickness 1.0mm, 99.97%
Bi
GF75246183
Bismuth, foil, 50x50mm, thickness 2.0mm, 100%
Bi
GF46489135
Bismuth, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF86892441
Bismuth, foil, light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF40931546
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF35246831
Bismuth, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF95571995
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF12550935
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF09655263
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF71673060
Bismuth, foil, light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF09816307
Bismuth, foil, light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF91742328
Bismuth, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF24083692
Bismuth, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF77293592
Bismuth, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF93912891
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF19216087
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi